Analytical standard, Cimaterol-d₇, reference material, Honeywell Fluka™
HONCR1022-10MG
:
- Pack type:Glass Bottle
- Pk:10 mg
- Standards:Analytical standard, neat
- CAS No.:1228182-44-2
- Description:Cimaterol-d₇
- Melting point:163…168 °C
- Formula:C₁₂D₇H₁₀N₃O
- Storage temperature:Ambient
- Description:Cimaterol-d₇
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results