Analytical standard, Chlorpromazine-d₆ hydrochloride, reference material, Honeywell Fluka™
HONCR1020-10MG
:
- Pack type:Glass Bottle
- Pk:10 mg
- Standards:Analytical standard, neat
- CAS No.:1228182-46-4
- Description:Chlorpromazine-d₆ hydrochloride
- Melting point:193…198 °C
- Formula:C₁₇D₆H₁₃ClN₂S·HCl
- Storage temperature:Ambient
- Description:Chlorpromazine-d₆ hydrochloride
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results