Analytical standard, Sulfaquinoxaline-phenyl-¹³C₆, reference material, Honeywell Fluka™
HONCR1016-10MG
:
- Pack type:Glass Bottle
- Pk:10 mg
- Standards:Analytical standard, neat
- CAS No.:1202864-52-5
- Description:Sulfaquinoxaline-phenyl-¹³C₆
- Melting point:263…267 °C
- Formula:C₈¹³C₆H₁₂N₄O₂S
- Storage temperature:Ambient
- Description:Sulfaquinoxaline-phenyl-¹³C₆
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results