Analytical standard, 3-Hydroxyibuprofen, reference material, Honeywell Fluka™
HONCR1013-10MG
:
- Pack type:Glass Bottle
- Pk:10 mg
- Standards:Analytical standard, neat
- CAS No.:53949-54-5
- Description:3-Hydroxyibuprofen
- Melting point:99 °C
- Formula:C₁₃H₁₈O₃
- Storage temperature:Ambient
- Description:3-Hydroxyibuprofen
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results