Order Entry
Czech Republic
ContactUsLinkComponent
 
Analytical standard, Phenylbutazon-¹³C₁₂, reference material, Honeywell Fluka™
  HONCR1012-10MG
 :  
Analytical standard, Phenylbutazon-¹³C₁₂, reference material, Honeywell Fluka™
  HONCR1012-10MG
 :  

 

  • Pack type:
    Glass Bottle
  • Pk:
    10 mg
  • Standards:
    Analytical standard, neat
  • CAS No.:
    1325559-13-4
  • Description:
    Phenylbutazon-¹³C₁₂
  • Melting point:
    109…110 °C
  • Formula:
    ¹³C₁₂C₇H₂₀N₂O₂
  • Storage temperature:
    Ambient
  • Description:
    Phenylbutazon-¹³C₁₂

 

 

  • High quality organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results