Analytical standard, Niflumic acid-(phenyl-¹³C₆), reference material, Honeywell Fluka™
HONCR1011-10MG
:
- Pack type:Glass Bottle
- Pk:10 mg
- Standards:Analytical standard, neat
- CAS No.:1325559-33-8
- Description:Niflumic acid-(phenyl-¹³C₆)
- Melting point:204 °C
- Formula:¹³C₆C₇H₉F₃N₂O₂
- Storage temperature:Ambient
- Description:Niflumic acid-(phenyl-¹³C₆)
- High quality organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results