HPLC columns, NUCLEODUR® PFP
Supplier: MACHEREY-NAGEL
Certificates
About this item
NUCLEODUR® PFP is a pentafluorophenyl-propyl modified silica phase for HPLC.
- High purity spherical silica gel
- Hydrophobic, polar RP phase with additional aromatic and steric selectivity
- Separation mechanism based on 4 retention mechanisms (polar interactions, dipole-dipole interactions, π-π interactions, hydrophobic interactions)
The separation mechanism features an alternative selectivity in comparison to classical C18 phases. Thus NUCLEODUR® PFP is well applicable for polar compounds like pharmaceuticals and phenols, halogenated compounds, aromatic and unsaturated compounds, isomers, and basic compounds - also for LC-MS detection.
EC guard columns are used for analytical EC columns with the Column Protection System guard column holder (554-5851).
Specifications
- Catalog No:
- MANA760446.30
- MANA760446.40
- MANA760446.46
- MANA760443.40
- MANA760443.46
- MANA760445.46
- MANA760446.20
- MANA760448.20
- MANA760448.30
- MANA760443.20
- MANA760448.40
- MANA760443.30
- MANA760448.46
- MANA760447.20
- MANA760447.30
- MANA760447.40
- MANA760447.46
- MANA760438.20
- MANA760436.20
- MANA760436.30
- MANA760436.40
- MANA760436.46
- MANA760433.30
- MANA760459.30
- MANA760433.40
- MANA760459.40
- MANA760459.46
- MANA760433.46
- MANA760435.20
- MANA760458.46
- MANA760433.20
- MANA760459.20
- MANA761977.30
- MANA761977.20
- MANA760458.40
- MANA760457.40
- MANA760457.46
- MANA760458.20
- MANA760458.30
- MANA760456.40
- MANA761976.20
- MANA760456.46
- MANA761976.30
- MANA760457.20
- MANA760457.30
- MANA760431.30
- MANA760431.40
- MANA760431.46
- MANA761975.20
- MANA761975.30
- MANA760456.30
- MANA760431.20
- MANA760455.46
- MANA760456.20
- MANA760453.20
- MANA760453.30
- MANA760453.40
- MANA760453.46
- MANA760449.20
- MANA760449.30
- MANA760449.40
- MANA760449.46
- Carbon load:
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- 8%
- Endcapped:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- pH range:
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- 1 - 9
- USP code:
- L43
- L43
- L43
- L43
- L43
- L43
- L43
- L43
- L43
- L43
- L43
- L43
- L43
- L43
- L43
- L43
- L43
- L43
- L43
- L43
- L43
- L43
- L43
- L43
- L43
- L43
- L43
- L43
- L43
- L43
- L43
- L43
- L43
- L43
- L43
- L43
- L43
- L43
- L43
- L43
- L43
- L43
- L43
- L43
- L43
- L43
- L43
- L43
- L43
- L43
- L43
- L43
- L43
- L43
- L43
- L43
- L43
- L43
- L43
- L43
- L43
- L43
Specifications
Guard column holders for VP columns
Product Information
Frequently Bought Together