Order Entry
Czech Republic
ContactUsLinkComponent
Fmoc-Asp(Ompe)-OH 98% (by HPLC)
Fmoc-Asp(Ompe)-OH 98% (by HPLC)
Catalog # H66281.06
CAS Number:  
Fmoc-Asp(Ompe)-OH 98% (by HPLC)
Catalog # H66281.06
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Assay (HPLC):
    97.5%

Specification Test Results

About this item

 :
 :439.508 g/mol
Storage Temperature:Freezer
CAS Number:180675-08-5
Formula:C₂₅H₂₉NO₆
MDL Number:MFCD10001333