Ethyl cellulose, (22 cP, 5 % in toluene/ethanol 80:20), Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:9004-57-3
:
MDL Number:MFCD00131037
Density:1.14 g/ml (25 °C) (lit.)
:M-Clarity™ quality level = MQ100
Specification Test Results
- Infrared spectrum:Conforms to Structure
- Ethoxyl (Assay):47.5 - 49.5%
- Viscosity (C=5%, Toluene:Ethanol 80:20 at 25 °C):18 - 24 cps
Specification Test Results
Frequently Bought Together