ContactUsLinkComponent
MTT cell proliferation assay kit
MTT cell proliferation assay kit
Catalog # OZBIMT01000
Supplier:  OZ BIOSCIENCES
CAS Number:  
undefined
MTT cell proliferation assay kit
Catalog # OZBIMT01000
Supplier:  OZ BIOSCIENCES
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    MTT cell proliferation assay kit, contains MTT reagent 10X (10×1 ml) and solubilisation solution (100 ml)
Specifications
About this item
MTT cell proliferation assay kit is designed for spectrophotometric quantification of cell growth, viability and proliferation and can be used as a direct indicator of cytotoxicity (such as for screening anticancer drugs) and apoptosis.
  • Easy and ready to use
  • Accurate, absorbance signal produced correlates with the cell number
  • Convenient packaging
  • Economical
Kit is a colorimetric assay for measuring the mitochondrial reductases activity in living cells. It is based on the cleavage of membrane-permeable yellow tetrazolium salt MTT to blue/purple formazan crystals by metabolically active cells. This MTT assay kit contains sufficient reagents to perform 1000 assays in 96-well plate format.
Caution: Store MTT Reagent at –20 °C MTT reagent and solubilisation solution at +4 °C.