

8-Mercaptomenthone cis- and trans-mixture (natural) ≥95%, FG Halal, Kosher, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:38462-22-5
:
Formula:C₁₀H₁₈OS
MDL Number:MFCD00012393
:186.31 g/mol
Boiling Pt:56 °C/0.1 mmHg (lit.)
Density:1 g/ml at 25 °C (lit.) (lit.)
Flash Pt:108 °C
UN:2810
ADR:6.1,III
:M-Clarity™ quality level = MQ300
:GHS06
:GHS09
Specification Test Results
- Assay:≥95%
- Optical Activity:[α]20/D −21°, neat
- Organoleptic :Berry; Tropical; Fruity; Minty; Sulphurous
- Food Allergen:No Known Allergens
- Fragrance Allergen:No Known Allergens
Specification Test Results
Frequently Bought Together