Order Entry
Czech Republic
ContactUsLinkComponent
Ethyl 4-hydroxybenzoate 98.0-102.0%, GenAR® NF, Macron Fine Chemicals™
Ethyl 4-hydroxybenzoate 98.0-102.0%, GenAR® NF, Macron Fine Chemicals™
Catalog # 6882-18
CAS Number:  
undefined
Ethyl 4-hydroxybenzoate 98.0-102.0%, GenAR® NF, Macron Fine Chemicals™
Catalog # 6882-18
CAS Number:  

Specifications

  • Pk:
    25 kg
  • Seq:
    4283

Specifications

Specification Test Results

  • Bulk Pharmaceutical Chemical:
    98.0 - 102.0%
  • NF - Assay (C₉H₁₀O₃):
    98.0 - 102.0 %
  • NF - Appearance (white, crystalline powder):
    Passes Test
  • NF - Color of Solution:
    Passes Test
  • NF - Identification A:
    Passes Test
  • NF - Identification B:
    Passes Test
  • NF - Acidity:
    Passes Test
  • NF - Related Substances - p-Hydroxybenzoic Acid:
    ≤0.5 %
  • NF - Related Substances - Unspecified Impurities (each):
    ≤0.5 %
  • NF - Related Substances - Total Impurities:
    ≤1.0 %
  • NF - Residue on Ignition:
    ≤0.1 %
  • EP/BP - Assay (C₉H₁₀O₃):
    98.0 - 102.0 %
  • EP/BP - Appearance of Solution:
    Passes Test
  • EP/BP - Identification A:
    Passes Test
  • EP/BP - Identification B:
    Passes Test
  • EP/BP - Acidity:
    Passes Test
  • EP/BP - Related Substances - Impurity A:
    ≤0.5 %
  • EP/BP - Related Substances - Unspecified Impurities (each):
    ≤0.5 %
  • EP/BP - Related Substances - Total Impurities:
    ≤1.0 %
  • EP/BP - Sulfated Ash:
    ≤0.1 %
  • JP - Acidity:
    Passes Test
  • JP - Assay:
    98.0 - 102.0 %
  • JP - Clarity and Color of Solution:
    Passes Test
  • JP - Identification:
    Passes Test
  • JP - Melting Point:
    115 - 118 °C
  • JP - Heavy Metals:
    ≤20 ppm
  • JP-RelatedSubstances- p-Hydroxybenzoic Acid:
    ≤0.5 %
  • JP-Related Substances-Unspecified Impurities (each):
    ≤0.5 %
  • JP-Related Substances-Total Impurities:
    ≤1.0 %
  • JP - Residue on Ignition:
    ≤0.10 %

Specification Test Results

About this item

 :
 :166.18 g/mol
CAS Number:120-47-8
Formula:C₉H₁₀O₃
MDL Number:MFCD00002353