Order Entry
Czech Republic
ContactUsLinkComponent
2-[2-(2-Aminoethoxy)ethoxy]ethanol ≥96% (by GC)
Corrosive
2-[2-(2-Aminoethoxy)ethoxy]ethanol ≥96% (by GC)
Supplier:  Merck
Certificates
Corrosive
2-[2-(2-Aminoethoxy)ethoxy]ethanol ≥96% (by GC)
Supplier:  Merck

Specification Test Results

  • APPEARANCE (COLOR):
    Colorless
  • APPEARANCE (FORM):
    Liquid
  • TITRATION (NT) HCLO4 0.1M:
    96.0 - 104.0 %
  • PURITY (GC AREA %):
    ≥ 96.0 %
  • INFRARED SPECTRUM:
    CONFORMS TO STRUCTURE

Specification Test Results

Frequently Bought Together