Organic reference standards, (±)-geosmin 100 µg/ml in methanol, Dr Ehrenstorfer
Catalog # EHERXA14005000ME
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pk:1 mL
- Standards:Reference standard
- CAS No.:16423-19-1
- Description:(±)-Geosmin 100 µg/ml in methanol
- Formula:C₁₂H₂₂O
- Description:(±)-Geosmin
Specifications
About this item
Certifications: Produced in accordance to ISO 17034