Zadání objednávky
Czech Republic
ContactUsLinkComponent
3,3'-Di-n-hexyloxacarbocyanine iodide [DiOC6(3) iodide] ≥98%, crystalline solid
Irritant
3,3'-Di-n-hexyloxacarbocyanine iodide [DiOC6(3) iodide] ≥98%, crystalline solid
Supplier:  Cayman Chemical
Certificates
Irritant
3,3'-Di-n-hexyloxacarbocyanine iodide [DiOC6(3) iodide] ≥98%, crystalline solid
Supplier:  Cayman Chemical

Specification Test Results

  • Purity:
    ≥98%
  • Emission:
    501 nm
  • Excitation:
    484 nm
  • λmax:
    486 nm

Specification Test Results

Frequently Bought Together