ContactUsLinkComponent
Thermal transfer printer labels, type B-7604
Thermal transfer printer labels, type B-7604
Catalog # BRDY236703
Supplier:  Brady
undefined
Thermal transfer printer labels, type B-7604
Catalog # BRDY236703
Supplier:  Brady
Specifications
  • Type:
    B-7604
  • Colour:
    White
  • Description:
    Printer labels
  • Dimensions:
    57×19 mm
  • Material:
    Paper
  • Seq:
    28
  • Model:
    BPT-522-075
  • Packed:
    21360 labels/box
  • Pk:
    21.360
  • Adhesive:
    Permanent acrylic
Specifications
About this item
Printable, paper labels for thermal transfer printers.