Protein delivery reagent, Ab-DeliverIN™
Supplier: OZ BIOSCIENCES
Certificates
About this item
Ab-DeliverIN™ transfection reagent is dedicated to the intracellular delivery of fully functional antibodies. It opens up new fields of investigation in proteomics, for instance to elucidate complex molecular mechanisms or to design new potential therapies.
- Intracellular delivery of functionally active antibody
- Highly efficient in many primary cells and cell lines
- Serum compatible
- Non toxic
- Straightforward protocol and ready to use reagent
Store Ab-DeliverIN™ at 4 °C and FITC labeled IgG at –20 °C.
Delivery: Shipped at room temperature.
Specifications
- Protein delivery:Yes