Transduction reagent, Viro-MICST™
Viro-MICST™ transduction reagent allows genetically modifying cells directly on magnetic cell purification columns. Based on i-MICST™ Technology (Integrated Magnetic Immuno-Cell Sorting and Transfection/Transduction), this technology combines cell isolation and genetic modification in one simple and reliable integrated system. For efficient and selective gene delivery to target cells, Viro-MICST™ can be used in combination with any viral vector and allows you to reduce cell manipulation steps and save time and material.
- Highly efficient
- Rapid, simple, and ready to use
- Suitable for all cells
Store at +4 °C.
: Shipped at room temperature.
- Cell specific:CD4+, CD8+, T lymphocytes
Frequently Bought Together