Order Entry
Czech Republic
ContactUsLinkComponent
Guanidinium hydrochloride 99.5+% for molecular biology
Irritant
Guanidinium hydrochloride 99.5+% for molecular biology
Certificates
undefined
Irritant
Guanidinium hydrochloride 99.5+% for molecular biology
Certificates

Specification Test Results

  • Form:
    White/clear crystals
  • Assay, Absorbance (225 nm, 6 M, 1 cm):
    ≤0.2
  • Assay, Absorbance (260 nm, 6 M, 1 cm):
    ≤0.03
  • Assay, Aldehyde:
    None detected
  • Assay, Ammeline:
    None detected
  • Assay:
    ≥99.5%
  • Impurity content, Sulfate (SO₄):
    ≤ 50 ppm
  • Melting Point:
    184 ±3°C
  • Solution Test, (6 M, H₂O):
    Clear and colourless
  • Identification (FTIR):
    Conforms
  • Dnase (endo):
    Passes Test
  • Protease Activity:
    Passes Test
  • RNase:
    Passes Test
  • Clear and colorless:
    ≤1 ppm
  • Trace Metal: Arsenic (As):
    Solution Test (6M, H2O)
  • Trace Metal: Copper (Cu):
    ≤1 ppm
  • Trace Metal: Iron (Fe):
    ≤2 ppm
  • Trace Metal: Lead (Pb):
    ≤5 ppm
  • Trace Metal: Zinc (Zn):
    ≤1 ppm

Specification Test Results

Related Information

Frequently Bought Together