
Dithiothreitol (DTT, Cleland's reagent) ≥99.5%, Ultrapure for molecular biology
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:3483-12-3
:27565-41-9
Formula:C₄H₁₀O₂S₂
MDL Number:MFCD00004877
:154.25 g/mol
Merck Index:14,03376
Boiling Pt:125…130 °C (12 mmHg)
Melting Pt:40…43 °C
Flash Pt:>110 °C (230 °F)
EINECS:222-468-7
:GHS07
Storage Temperature:Freezer
Specification Test Results
- Assay (SH):≥99.5%
- Assay Absorbance (260 nm, 0.1 M, 1 cm):≤0.40
- Assay Absorbance (280 nm, 0.1 M, 1 cm):≤0.08
- DNase:Passes Test
- Impurity Content, OxidiSed DTT:≤0.3%
- Protease Activity:Passes Test
- RNase:Passes Test
Specification Test Results
Frequently Bought Together