Order Entry
Czech Republic
ContactUsLinkComponent
 
 
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) 99.8-100.1% (dry basis), Ultrapure for molecular biology
 
undefined
 
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) 99.8-100.1% (dry basis), Ultrapure for molecular biology
 

 

  • Assay (unspecified) - Dry Basis:
    99.8 - 100.1%
  • Assay (unspecified) - Absorbance (250 nm, 1 M, 1 cm):
    ≤0.1
  • Assay (unspecified) - Absorbance (290 nm, 40%, 1 cm):
    ≤0.2
  • Identification (FTIR):
    Conforms
  • Residue on Ignition:
    ≤0.1%
  • Insoluble Material:
    ≤0.005%
  • Water Content (Karl Fischer Titration):
    ≤1%
  • Loss on Drying:
    ≤1%
  • pH (5%, H₂O):
    10.0 - 11.5
  • pKa (20 °C):
    8.30 ±0.15
  • Arsenic (As):
    ≤1 ppm
  • Barium (Ba):
    ≤1 ppm
  • Cadmium (Cd):
    ≤1 ppm
  • Calcium (Ca):
    ≤1 ppm
  • Iron (Fe):
    ≤1 ppm
  • Lead (Pb):
    ≤1 ppm
  • Magnesium (Mg):
    ≤1 ppm
  • Manganese (Mn):
    ≤1 ppm
  • Zinc (Zn):
    ≤1 ppm

 

Frequently Bought Together