Order Entry
Czech Republic
ContactUsLinkComponent
 
 
L(+)-Cysteine hydrochloride monohydrate 98.5-101.5% (dry basis)
 
undefined
 
L(+)-Cysteine hydrochloride monohydrate 98.5-101.5% (dry basis)

 

  • Form:
    White crystalline powder
  • Assay (unspecified):
    98.5 to 101.5% (dry basis)
  • Identification (FTIR):
    Conforms
  • Residual halide: Chlorine (Cl):
    19.8 - 20.8%
  • Residue on ignition:
    ≤ 0.1%
  • Impurity content: Sulfate (SO4):
    ≤ 0.03%
  • Loss on Drying:
    8.0 to 12.0%
  • Optical Rotation:
    +5.7° to 6.8° (c=8 in in 6N HCl, dry basis)
  • Thin Layer Chromatography:
    Homogeneous
  • Trace Metal: Arsenic (As):
    ≤ 1.5 ppm
  • Trace Metal: Heavy Metals (Pb):
    ≤ 10 ppm
  • Trace Metal: Iron (Fe):
    ≤ 10 ppm

 

Frequently Bought Together