ContactUsLinkComponent
Liquid chromatography sorbents, Aluminium Oxide 90
Liquid chromatography sorbents, Aluminium Oxide 90
Supplier:  MACHEREY-NAGEL
Certificates
undefined
Liquid chromatography sorbents, Aluminium Oxide 90
Supplier:  MACHEREY-NAGEL
Certificates
Specifications
  • Type:
    Aluminium oxide 90 acidic
  • Working pH:
    pH 4 ± 0,3