Deep well V-bottom plates, 24-well
SARPA-0038
- Packed:Individually wrapped
- Sterile:+
- Colour:Amber
- Description:Deep 24-well V-bottom plates
- Environmentally Preferable:
- Pk:1
- Well volume:8 ml
These 24-well deep well plates can be used to hold samples harvested from bioreactors. They can also be used to hold reagents and feeds on the storage deck, before being added into the bioreactor.
- Irradiated
: Pack contains 25 sterile plates with lids, individually wrapped.