Order Entry
Czech Republic
ContactUsLinkComponent
Extractor, for light solvents
Extractor, for light solvents
Catalog # WINZ21-550-81
Extractor, for light solvents
Catalog # WINZ21-550-81

Specifications

  • Extraction cap.:
    1000 ml
  • Socket size:
    71/51 NS
  • Description:
    Extractor with valve stopcock, cone NS 29/32
  • Pk:
    1

Specifications

About this item

Made of borosilicate glass. Extractor specially designed to extract liquids for light solvents.

  • Very good chemical resistance
  • Very good temperature resistance

This unit is designed with valve stopcock with 0,3 mm PTFE spindle.