0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
Standards
Cationic Standards
Iron Standards
Iron Standard in Oil, 1000 ppm, Conostan
Iron Standard in Oil, 1000 ppm, Conostan
Catalog # PROOCON150-100-265
Dodavatel:
LGC Standards PROMOCHEM
Číslo CAS:
Expand image
Iron Standard in Oil, 1000 ppm, Conostan
Catalog # PROOCON150-100-265
Dodavatel:
LGC Standards PROMOCHEM
Číslo CAS:
Specifikace
Concentration:
1000 µg/g
Matrix:
oil
Pk:
50 g
Element:
Iron
Symbol:
Fe
Specifikace
O tomto produktu
Product Details
Product Details