Dehydrated culture media, for microbiology
Catalog # APPC413812.1210
Supplier: PanReac AppliChem
CAS Number:
Specifications
- Pk:500 g
- Description:Slanetz bartley medium
- Food:+
Specifications
About this item
Culture medium for the detection and enumeration of Enterococci in water and foods.
- pH: 7,2 ±0,2
- Soluble in water
- Store in a cool and dry place
Medium contains yeast extract (5,0 g/l); D(+)-glucose (2,0 g/l); sodium azide (0,4 g/l); dipotassium hydrogen phosphate (4,0 g/l); 2,3,5-triphenyl-2H-tetrazolium chloride (0,1 g/l); tryptose (20,0 g/l) and agar (10,0 g/l).