Order Entry
Czech Republic
ContactUsLinkComponent
Organic reference standard, Azoxystrobin, Dr. Ehrenstorfer
Organic reference standard, Azoxystrobin, Dr. Ehrenstorfer
Catalog # EHERL10413000CY
Supplier:  EHRENSTORFER
CAS Number:  
Organic reference standard, Azoxystrobin, Dr. Ehrenstorfer
Catalog # EHERL10413000CY
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pk:
    10 mL
  • Standards:
    Reference standard
  • CAS No.:
    131860-33-8
  • Description:
    Azoxystrobin, 10 μg/ml in cyclohexane
  • Formula:
    C₂₂H₁₇N₃O₅
  • Description:
    Azoxystrobin

Specifications

About this item

Certifications: Accredited by ISO/IEC 17025.