ESD safety ankle boots, slip-on, Chelsea XW
: ELTEN
Comfortable ESD Chelsea boots made from cowhide leather plus a mono-density PU sole (Business).
- Light leather lining
- Replaceable insole; orthopaedic insoles possible
- Full length inlay sole (Business ESD)
- Soft fleece ESD insole
- Protective steel toecap (200 J)
- Extra wide version (XW)
Ideal for use in areas where exposure to moisture is expected.
Colour: Black
: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S2, SRC.
Frequently Bought Together