Order Entry
Czech Republic
ContactUsLinkComponent
Sodium hydroxide 30% in aqueous solution, TECHNICAL
 
Sodium hydroxide 30% in aqueous solution, TECHNICAL
  VWRS28217.980
 :  
 
Sodium hydroxide 30% in aqueous solution, TECHNICAL
  VWRS28217.980
 :  

 

  • Pk:
    1.000 L
  • Seq:
    5037

 

 

  • Assay:
    Min. 29 %
  • Heavy metals (as Pb):
    Max. 50 ppm

 

 

 :
 :40 g/mol
 :119…135 °C (1013 hPa)
 :9…10 °C
 :Ambient
 :215-185-5
 :1824
 :8,II
 :GHS05
 :1310-73-2
 :NaOH
 :MFCD00003548