

Potassium hydroxide 0.5 mol/l (0.5 N) in ethanol, AVS TITRINORM volumetric solution
Catalog # VWRS31308.370
Supplier: VWR CUSTOM MADE CHEMICALS
CAS Number:
Specifications
- Pk:5 L
- Seq:5190
Specifications
Specification Test Results
- Titer (20°C; real value 0.2 % accuracy):0.499 - 0.501 mol/l
- NIST traceable:Confirmed
Specification Test Results
About this item
:
:56.11 g/mol
Storage Temperature:Ambient
EINECS:215-181-3
UN:2924
ADR:3 (8),II
:GHS02
:GHS05
CAS Number:1310-58-3
Formula:KOH
MDL Number:MFCD00003553