Hydrochloric acid 4 mol/l (4 N), VOLUSOL® volumetric solution
Catalog # VWRS310701.9025
Supplier: VWR Collection
CAS Number:
Specifications
- Pack type:Plastic container
- Pk:25 L
- Seq:5327
Specifications
Specification Test Results
- Titer:3.984 - 4.016 mol/l
- NIST traceable:Confirmed
Specification Test Results
About this item
:
Density:1.06 g/cm³ (20 °C)
EINECS:231-595-7
ADR:8,III
:36.46 g/mol
UN:1789
:GHS07
:GHS05
Storage Temperature:Ambient
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324