
Triammonium citrate 97-103%, GPR RECTAPUR®
27153.460
:
- Pk:25 kg
- Seq:4283
- Assay:97 - 103 %
- Identification:Passes test
- pH (20°C; 5 %):6.0 - 7.5
- Ignition residue (SO4):Max. 0.05 %
- Cl (Chloride):Max. 100 ppm
- Pb (Lead):Max. 10 ppm
:
:243.22 g/mol
:185 °C
:1 g/cm³ (25 °C)
:222-394-5
:GHS07
:Ambient
:3458-72-8
:C₆H₁₇N₃O₇
:MFCD00036406