

Ethyl acetate ≥99%, TECHNICAL
VWRS23879.410
:
- Pack type:Plastic container
- Pk:10 L
- Seq:5214
- Assay:Min. 99 %
- Appearance:Clear colourless liquid
- IR Spectrum:Passes test
:
:88.11 g/mol
:13,03792
:77.1 °C (1013 hPa)
:–83 °C
:0.902 g/cm³ (20 °C)
:–4 °C
:Ambient
:205-500-4
:1173
:3,II
:01-2119475103-46
:GHS02
:GHS07
:141-78-6
:CH₃COOC₂H₅
:MFCD00009171