Order Entry
Czech Republic
ContactUsLinkComponent
Aluminium nitrate nonahydrate ≥98.0% analytical reagent
Aluminium nitrate nonahydrate ≥98.0% analytical reagent
Catalog # 21050.460
Supplier:  VWR Collection
CAS Number:  
Aluminium nitrate nonahydrate ≥98.0% analytical reagent
Catalog # 21050.460
Supplier:  VWR Collection
CAS Number:  

Specifications

  • Pk:
    25 kg
  • Seq:
    4283

Specifications

Specification Test Results

  • Assay:
    Min. 98.0 %
  • Heavy metals (as Pb):
    Max. 10 ppm
  • Insolubility in water:
    Max. 50 ppm
  • Not precipitated by NH4OH (as SO4):
    Max. 0.1 %
  • Cl (Chloride):
    Max. 10 ppm
  • SO4 (Sulphate):
    Max. 100 ppm
  • Fe (Iron):
    Max. 50 ppm

Specification Test Results

About this item

 :
 :375.13 g/mol
Melting Pt:73 °C
Density:1.72 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:236-751-8
UN:1438
ADR:5.1,III
 :GHS03
 :GHS07
CAS Number:7784-27-2
Formula:Al(NO₃)₃·9H₂O
MDL Number:MFCD00003419