Order Entry
Czech Republic
ContactUsLinkComponent
Sodium hydroxide 1.2 mol/l (1.2 N) in aqueous solution
Corrosive
Sodium hydroxide 1.2 mol/l (1.2 N) in aqueous solution
Catalog # VWRS5329.9010
Supplier:  VWR Collection
CAS Number:  
Corrosive
Sodium hydroxide 1.2 mol/l (1.2 N) in aqueous solution
Catalog # VWRS5329.9010
Supplier:  VWR Collection
CAS Number:  

Specifications

  • Pack type:
    Plastic container
  • Pk:
    10 L
  • Seq:
    5214

Specifications

Specification Test Results

  • Titer:
    1.185 - 1.215 mol/l
  • NIST traceable:
    Confirmed

Specification Test Results

About this item

 :
 :40 g/mol
Density:1.04 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:215-185-5
UN:1824
ADR:8,II
 :GHS05
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548