Cyclohexane ≥99.5%, AnalaR NORMAPUR® ACS, Reag. Ph. Eur. analytical reagent
Catalog # 23224.360
Supplier: VWR Collection
CAS Number:
Specifications
- Pack type:Plastic container
- Pk:5 L
- Seq:5190
Specifications
Specification Test Results
- Assay (on anhydrous substance):Min. 99.5 %
- IR Spectrum:Passes test
- Readily carbonisable substances:Passes test
- Boiling point:79 - 81 °C
- Colouration:Max. 10 APHA
- Density (20/4):0.778 - 0.779
- Density (20/20):0.779 - 0.780
- Free acidity:Max. 0.0003 meq/g
- Solidification point:5.5 - 6.5 °C
- Substances coloured by H2SO4:Max. 60 APHA
- Benzene:Max. 100 ppm
- Cyclohexene:Max. 100 ppm
- Evaporation residue:Max. 10 ppm
- Water:Max. 100 ppm
- Al (Aluminium):Max. 0.1 ppm
- Ba (Barium):Max. 0.02 ppm
- Ca (Calcium):Max. 0.5 ppm
- Cd (Cadmium):Max. 0.05 ppm
- Co (Cobalt):Max. 0.02 ppm
- Cr (Chromium):Max. 0.02 ppm
- Fe (Iron):Max. 0.1 ppm
- K (Potassium):Max. 0.1 ppm
- Mg (Magnesium):Max. 0.05 ppm
- Mn (Manganese):Max. 0.02 ppm
- Na (Sodium):Max. 1 ppm
- Ni (Nickel):Max. 0.02 ppm
- Pb (Lead):Max. 0.05 ppm
- Sn (Tin):Max. 0.1 ppm
- Sr (Strontium):Max. 0.02 ppm
- Zn (Zinc):Max. 0.1 ppm
- Conforms to ACS:Passes test
- Conforms to Reag. Ph.Eur.:Passes test
Specification Test Results
About this item
:
:84.16 g/mol
Merck Index:13,02752
Boiling Pt:81 °C (1013 hPa)
Melting Pt:7 °C
Density:0.7785 g/cm³ (20 °C)
Flash Pt:–18 °C
Storage Temperature:Ambient
EINECS:203-806-2
UN:1145
ADR:3,II
REACH:01-2119463273-41
:GHS02
:GHS08
:GHS07
:GHS09
CAS Number:110-82-7
Formula:C₆H₁₂
MDL Number:MFCD00003814