Order Entry
Czech Republic
ContactUsLinkComponent
Sodium hydroxide 50% in aqueous solution, GPR RECTAPUR®
Corrosive
Sodium hydroxide 50% in aqueous solution, GPR RECTAPUR®
Catalog # VWRS84522.550
Supplier:  VWR Collection
CAS Number:  
Corrosive
Sodium hydroxide 50% in aqueous solution, GPR RECTAPUR®
Catalog # VWRS84522.550
Supplier:  VWR Collection
CAS Number:  

Specifications

  • Pack type:
    Plastic drum
  • Pk:
    200 L
  • Seq:
    5549

Specifications

Specification Test Results

  • Assay:
    47 - 51 %
  • Sodium carbonate:
    Max. 0.5 %
  • Heavy metals (as Pb):
    Max. 20 ppm
  • Cl (Chloride):
    Max. 50 ppm
  • Fe (Iron):
    Max. 50 ppm

Specification Test Results

About this item

 :
 :40 g/mol
Boiling Pt:119…135 °C (1013 hPa)
Melting Pt:9…10 °C
Density:1.53 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-185-5
UN:1824
ADR:8,II
 :GHS05
CAS Number:1310-73-2
Formula:NaOH
MDL Number:MFCD00003548