Order Entry
Czech Republic
ContactUsLinkComponent
Potassium bromide ≥98.5%, GPR RECTAPUR®
Potassium bromide ≥98.5%, GPR RECTAPUR®
Catalog # 26704.361
Supplier:  VWR Collection
CAS Number:  
Potassium bromide ≥98.5%, GPR RECTAPUR®
Catalog # 26704.361
Supplier:  VWR Collection
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    5 kg
  • Seq:
    4114

Specifications

Specification Test Results

  • Assay:
    Min. 98.5 %
  • Heavy metals (as Pb):
    Max. 10 ppm
  • Cl (Chloride):
    Max. 0.5 %
  • SO4 (Sulphate):
    Max. 100 ppm
  • Fe (Iron):
    Max. 10 ppm

Specification Test Results

About this item

 :
Merck Index:13,07701
Boiling Pt:1435 °C (1013 hPa)
Melting Pt:730 °C
Density:2.75 g/cm³ (20 °C)
 :119 g/mol
Storage Temperature:Ambient
EINECS:231-830-3
 :GHS07
CAS Number:7758-02-3
Formula:KBr
MDL Number:MFCD00011358