Order Entry
Czech Republic
ContactUsLinkComponent
 
Canister pumps
Catalog # 148-0168
Supplier:  Dr. Weigert
undefined
Canister pumps
Catalog # 148-0168
Supplier:  Dr. Weigert

Specifications

  • Description:
    Dispenser for 5 L, 10 L and 12 kg canisters (30 ml stroke)
  • Seq:
    01
  • Pk:
    1

Specifications

About this item