Orders LinkContactUsLinkComponent
Precision cut thermometers
Precision cut thermometers
Precision cut thermometers
These standard grinding size precision cut thermometers are designed with handle.
  • Colourless, round capillary form
Upper part: 275×11 mm
Lower part: Ø 7,25 mm
Product Family Options
  • Description
    Range
    Division
    Filling
    Immersion depth
    For NS
  • Uncovered type, with blank round capillary form; Installation length: 210 cm
    −20...+250 °C
    1,0 °C
    Blue
    184 mm
    19/26
    1 * 1 KS
    1 * 1 KS - 1 720,00 Kč
    1 720,00 Kč
    1 * 1 KS
       1 720,00 Kč
  • Uncovered type, with blank round capillary form; Installation length: 225 cm
    −20...+250 °C
    1,0 °C
    Blue
    199 mm
    19/26
    1 * 1 KS
    1 * 1 KS - 1 720,00 Kč
    1 720,00 Kč
    1 * 1 KS
       1 720,00 Kč
  • Uncovered type, with blank round capillary form; Installation length: 235 cm
    −20...+250 °C
    1,0 °C
    Blue
    209 mm
    19/26
    1 * 1 KS
    1 * 1 KS - 1 720,00 Kč
    1 720,00 Kč
    1 * 1 KS
       1 720,00 Kč