ContactUsLinkComponent
Affinity chromatography columns, HiScreen™ IXSelect
Catalog # GEHE17-3714-10
Supplier:  Cytiva
Affinity chromatography columns, HiScreen™ IXSelect
Catalog # GEHE17-3714-10
Supplier:  Cytiva
Specifications
  • Capacity:
    4,7 ml
  • Description:
    HiScreen™ IXSelect
  • Pk:
    1
Specifications
About this item
HiScreen™ IXSelect columns are prepacked with IXSelect affinity chromatography medium. These columns are an excellent choice for method optimisation and parameter screening.
  • Easily connected in series to achieve 20 cm bed height
  • Small bed volume gives fast results and minimal sample/buffer consumption
  • Reproducible results, scalable to BioProcess columns packed with the same chromatography resin using the same linear fluid velocity
IXSelect is a high-throughput resin specifically designed for the purification of factor IX targets from recombinant feed streams.