HPLC columns, Cogent C8 300™
Supplier: MICROSOLV
Certificates
About this item
Using proprietary bonding technology, the Cogent C8 300™ has a C8 chain bonded directly to the 5 µm, 300A TYPE-C™ silica surface with two separate points of attachment resulting in silcon-carbon bonds.
- Extremely long column life
- Polar and non-polar analytes
- Increase precision of your results
- Based on silica hydride
- Extremely low pH is possible with this column
The bonded does not hydrolyse in acidic conditions since there are no siloxane bonds attaching the ligands. This column is a very stable, efficient and unique HPLC column for macro molecules. A very powerful tool for bio separations labs working with large proteins, peptides, polysaccharides or tryptic digests looking for longer column life and improved precision in results.
Specifications
- Catalog No:
- 554-6158
- 554-6119
- 554-6160
- 554-6155
- 554-6116
- 554-6154
- 554-6115
- 554-6157
- 554-6118
- 554-6162
- 554-6156
- 554-6117
- 554-6161
- 554-6140
- 554-6112
- 554-6111
- 554-6159
- 554-6152
- 554-6440
- 554-6437
- 554-6110
- 554-6142
- 554-6441
- 554-6444
- 554-6445
- 554-6109
- 554-6108
- 554-6105
- 554-6104
- 554-6151
- 554-6150
- 554-6122
- 554-6103
- 554-6102
- 554-6149
- 554-6126
- Carbon load:
- 13 - 14%
- 5%
- 13 - 14%
- 13 - 14%
- 7%
- 13 - 14%
- 7%
- 13 - 14%
- 5%
- 13 - 14%
- 13 - 14%
- 7%
- 13 - 14%
- 5%
- 5%
- 13 - 14%
- 13 - 14%
- 11 - 12%
- 11 - 12%
- 7%
- 11 - 12%
- 11 - 12%
- 11 - 12%
- 7%
- 7%
- 5%
- 7%
- 13 - 14%
- 13 - 14%
- 13 - 14%
- 7%
- 7%
- 13 - 14%
- 5%
- Endcapped:
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- No
- pH range:
- 2,0 - 8,0
- 1,0 - 10,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 10,1
- 2,0 - 8,0
- 2,0 - 10,1
- 2,0 - 8,0
- 1,0 - 10,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 10,1
- 2,0 - 8,0
- 1,0 - 10,0
- 1,0 - 10,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 10,1
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 10,1
- 2,0 - 10,1
- 1,0 - 10,0
- 2,0 - 10,1
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 8,0
- 2,0 - 10,1
- 2,0 - 10,1
- 2,0 - 8,0
- 1,0 - 10,0
- Surface area:
- 390 m2/gm
- 150 m2/gm
- 390 m2/gm
- 390 m2/gm
- 350 m2/gm
- 390 m2/gm
- 350 m2/gm
- 390 m2/gm
- 150 m2/gm
- 390 m2/gm
- 390 m2/gm
- 350 m2/gm
- 390 m2/gm
- 150 m2/gm
- 150 m2/gm
- 390 m2/gm
- 390 m2/gm
- 340 m2/gm
- 340 m2/gm
- 350 m2/gm
- 340 m2/gm
- 340 m2/gm
- 340 m2/gm
- 350 m2/gm
- 350 m2/gm
- 150 m2/gm
- 350 m2/gm
- 390 m2/gm
- 390 m2/gm
- 390 m2/gm
- 350 m2/gm
- 350 m2/gm
- 390 m2/gm
- 150 m2/gm
Specifications
Frequently Bought Together