Order Entry
Czech Republic
ContactUsLinkComponent
Petroleum spirit, 40…60 °C, ChromAR® for HPLC, Macron Fine Chemicals™
Flammable
Irritant
Health
Environment
Petroleum spirit, 40…60 °C, ChromAR® for HPLC, Macron Fine Chemicals™
Catalog # 6756-25
CAS Number:  
undefined
Flammable
Irritant
Health
Environment
Petroleum spirit, 40…60 °C, ChromAR® for HPLC, Macron Fine Chemicals™
Catalog # 6756-25
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L
  • Seq:
    5096

Specifications

Specification Test Results

  • Residue after Evaporation:
    max. 0.0005 %
  • Titrable Acid (meq/g):
    max. 0.0005
  • Water (KF):
    max. 0.01 %
  • at 210 nm:
    max. 0.2
  • at 220 nm:
    max. 0.1
  • at 230 nm:
    max. 0.05
  • at 240 nm:
    max. 0.01
  • at 250 nm:
    max. 0.004

Specification Test Results

About this item

 :
Boiling Pt:40…60 °C
Flash Pt:–18 °C
Storage Temperature:Ambient
UN:1268
ADR:3,II
 :GHS02
 :GHS07
 :GHS08
 :GHS09
CAS Number:64742-49-0
 :64742-82-1
MDL Number:MFCD00081849