

Pyridine ≥99.5%, ChromAR® for HPLC, Macron Fine Chemicals™
Catalog # 6730-25
Supplier: MACRON AVANTOR BRAND
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:2,5 L
- Seq:5096
Specifications
Specification Test Results
- Assay (GC):min. 99.5 %
- Residue after Evaporation:max. 0.005 %
- Water (KF):max. 0.05 %
- at 340 nm:max. 0.07
- at 350 nm:max. 0.02
- at 400 nm:max. 0.01
Specification Test Results
About this item
:
Boiling Pt:115.5 °C
Density:0.98
Flash Pt:17 °C
Storage Temperature:Ambient
ADR:3,
:79.1 g/mol
UN:1486
:GHS02
:GHS07
CAS Number:110-86-1
Formula:C₅H₅N
MDL Number:MFCD00011732