ContactUsLinkComponent
Coated slides
Coated slides
Catalog # SIAL703176-10PAK
Supplier:  Merck
Coated slides
Catalog # SIAL703176-10PAK
Supplier:  Merck
Specifications
  • W×D:
    25×25 mm
  • Description:
    Indium tin oxide coated slides
  • Pk:
    10
Specifications
About this item
Indium tin oxide coated glass slides.
  • Good resistivity of 70-100 Ω/sq
  • Unpolished surface with 86 % transmittance