ContactUsLinkComponent
Wash bottles, integral, wide neck, Azlon®
Wash bottles, integral, wide neck, Azlon®
Catalog # 215-4162
Supplier:  SciLabware
undefined
Wash bottles, integral, wide neck, Azlon®
Catalog # 215-4162
Supplier:  SciLabware
Specifications
  • Colour code:
    Assorted colours
  • Capacity:
    500 ml
  • Pk:
    5
Specifications
About this item
Wide neck wash bottles with red, yellow, blue, green and orange PP caps.
  • Fitted with colour coded one-piece PP closure that maintains it's shape in use
  • Designed with integral spout to prevent contamination
  • Good chemical resistance
The round iIntegral wash bottle is moulded with an integral spout so you can say goodbye to the contamination caused by the inner tube touches the benchtop during re-filling.