Order Entry
Czech Republic
ContactUsLinkComponent
 
Human Recombinant GACRP-30 (from E. coli)
 
undefined
Human Recombinant GACRP-30 (from E. coli)

 

  • Conjugation:
    Unconjugated
  • Protein function:
    Cytokine
  • Protein/peptide type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage conditions:
    –20 °C
  • Endotoxin content:
    Endotoxin LAL (EU/µg) ≤1
  • Biological activity:
    Bioactivity Assay: Inhibition of M1 cell proliferation, ED₅₀ ≤2000 ng/ml, Bioactivity ≥500 units/mg
  • Gene ID:
    Q15848
  • Reconstitution instructions:
    Sterile 10 mM sodium phosphate, 0,5 mM DTT, pH 7,5 at 0,1 mg/ml
  • Endotoxin-free:
    N
  • Carrier-free:
    Y
  • Protease-free:
    N
  • Animal-free:
    N
  • Protein synonyms:
    adipolean|Adiponectin|apm-1
  • Protein/peptide name:
    GACRP-30
  • Purity:
    ≥90%
  • Molecular weight:
    16,7 kDa
  • Sequence:
    MKGEPGEGAY VYRSAFSVGL ETYVTIPNMP IRFTKIFYNQ QNHYDGSTGK FHCNIPGLYY FAYHITVYMK DVKVSLFKKD KAMLFTYDQY QENNVDQASG SVLLHLEVGD QVWLQVYGEG ERNGLYADND NDSTFTGFLL YHDTN
  • Endotoxin level:
    Low
  • Formulation:
    10 mM Sodium phosphate, 0,5 mM DTT, pH 7,5
  • Nuclease-free:
    N
  • Grade:
    RUO
  • Shipping temperature:
    RT
  • Tested applications:
    Bioassay

 

Frequently Bought Together