Human Recombinant IL-17F (from E. coli)
Interleukin 17F (IL-17F), a member of the IL-17 cytokine family, is secreted by activated CD4+ T cells and monocytes. IL-17F binds the IL-17 receptor related molecule, IL17RC, to promote the production of the interleukin 6 (IL-6), interleukin 8 (IL-8), and granulocyte macrophage colony-stimulating factor (GM-CSF) cytokines. IL-17F also functions to regulate matrix turnover rates, inhibit endothelial cell angiogenesis, and induce the endothelial cell expression of interleukin 2 (IL-2), monocyte chemoattractant protein-1 (MCP-1), and transforming growth factor beta 1 (TGF-b1).
- High quality
- Low endotoxin
- Affordable
: For research use only.
- Conjugation:Unconjugated
- Protein function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0,1
- Biological activity:Bioactivity Assay: Production of IL-6 from mouse NIH 3T3 cells, ED₅₀ ≤100 ng/ml, Bioactivity ≥1,0×10⁴ units/mg
- Gene ID:Q96PD4
- Reconstitution instructions:Sterile water at 0,1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein/peptide name:IL-17F
- Purity:≥95%
- Molecular weight:15,0/30,1 kDa
- Sequence:MRKIPKVGHT FFQKPESCPP VPGGSMKLDI GIINENQRVS MSRNIESRST SPWNYTVTWD PNRYPSEVVQ AQCRNLGCIN AQGKEDISMN SVPIQQETLV VRRKHQGCSV SFQLEKVLVT VGCTCVTPVI HHVQ
- Endotoxin level:Low
- Formulation:0,1% Trifluoroacetic acid (TFA)
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
- Tested applications:Bioassay
Frequently Bought Together