Human Recombinant IL-32alpha (from E. coli)
Supplier: Shenandoah Biotechnology
Certificates
About this item
Interleukin 32 alpha (IL-32 α) is one of six known splice variants of the IL-32 gene. IL-32 α induces the macrophage production of inflammatory cytokines, such as interleukin 8 (IL-8), tumor necrosis factor-alpha (TNF-α), and macrophage inflammatory protein 2 (MIP-2). IL-32 α expression is increased after the activation of T cells, natural killer (NK) cells, and interferon gamma-treated epithelial cells.
- High Quality
- Low Endotoxin
- Affordable
A COA is supplied with each product shipped detailing purity, formulation, biological activity, endotoxin levels, reconstitution instructions and recommended storage conditions. This product is for research use only and is not for use in humans.
Caution: For research use only.
Specifications
- Conjugation:Unconjugated
- Protein function:Cytokine
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:–20 °C
- Endotoxin content:Endotoxin LAL (EU/µg) ≤0,1
- Biological activity:Bioactivity: No biological activity data is available at this time
- Gene ID:P24001-4
- Reconstitution instructions:Sterile water at 0.1 mg/ml
- Endotoxin-free:N
- Carrier-free:Y
- Protease-free:N
- Animal-free:N
- Protein synonyms:NK4
- Protein/peptide name:IL-32alpha
- Purity:≥95%
- Molecular weight:14,9 kDa
- Sequence:MCFPKVLSDD MKKLKARMHQ AIERFYDKMQ NAESGRGQVM SSLAELEDDF KEGYLETVAA YYEEQHPELT PLLEKERDGL RCRGNRSPVP DVEDPATEEP GESFCDKSYG APRGDKEELT PQKCSEPQSS K
- Endotoxin level:Low
- Formulation:50 mM sodium phosphate, pH 7,5
- Nuclease-free:N
- Grade:RUO
- Shipping temperature:RT
Specifications
Frequently Bought Together